Use the Define Modification List feature to select which modifications are visible in the Modifications lists in the Assign Variable Modifications pane in the Oligonucleotide Sequence Editor. You can also select which modifications are included in the default sublist for quick loading.
Prerequisites
- You are viewing the Oligonucleotide Sequence Editor.
Procedure
- In the Oligonucleotide Sequence Editor, select Define Modification List on the command bar.
- The Define Modification List table opens, displaying the complete list of modifications supported by the application.
- The modifications listed in the Define Modification List table are color-coded as described in the table below.
Color | Description | Modifications lists status | Sublist for quick-loading status |
---|---|---|---|
Blue (bold) | Recommended default modifications | By default, visible in Modifications list | By default, selected for sublist for quick-loading |
Black | Other modifications | By default, visible in Modifications list | By default, not selected for sublist for quick-loading |
Purple (bold) | User-defined custom modifications | By default, visible in Modifications list | By default, not selected for sublist for quick-loading |
- In the Display Variable Modification column, select/deselect the checkboxes for modifications to add/remove them from the visible Modifications lists in the Assign Variable Modifications pane.
- In the Select Default Modification column, select/deselect the checkboxes for modifications to add/remove them from the default sublist for quick-loading.
- Select the Save button to save changes and close the window or select the Close button to close the window without saving any changes.