The following table lists the one-letter codes for amino acids used by the BioPharma Finder application.
NOTE
The one-letter amino acid codes are not case-sensitive.
One-letter code | Amino acid |
---|---|
A | alanine |
R | arginine |
N | asparagine |
D | aspartic acid |
C | cysteine |
E | glutamic acid |
Q | glutamine |
G | glycine |
H | histidine |
I | isoleucine |
L | leucine |
K | lysine |
M | methionine |
F | phenylalanine |
P | proline |
S | serine |
T | threonine |
W | tryptophan |
Y | tyrosine |
V | valine |