When you select the Perform Digestion checkbox at the top right of the Enter Peptide or Protein Sequence area of the Theoretical Protein/Peptide Manager, the application activates the Digestion Parameters tab where you can specify the protease, peptide length constraints, and number of missed cleavage sites allowed for the theoretical digestion.
Prerequisites
- You are viewing the Theoretical Protein/Peptide Manager.
- You selected the Perform Digestion checkbox.
Procedure
- Select the Digestion Parameters tab in the Theoretical Protein/Peptide Manager.
Figure Digestion Parameters (optional) pane - In the Select Protease area, select a protease to apply to the digestion.
- The activities at the N-terminal and C-terminal for the selected protease appear in the N-Term and C-Term boxes, but are not editable.
TIP
The list of proteases in the Theoretical Protein/Peptide Manager corresponds to the proteases available in the Peptide Mapping Analysis workflow.
To add a new protease or edit protease parameters, use the Identification page of the processing method editing wizard in the Peptide Mapping Analysis workflow.
- In the Peptide Length Constraints boxes, specify the lower and upper values for the peptide length range.
- In the Number of Allowed Missed Cleavage Sites box, specify the maximum number of internal cleavage sites within a peptide fragment that a protease can miss during the digestion process.