The following table describes the information in the All Possible Proteoforms table.
Table All Possible Proteoforms table information
Column
Description
Row number (bolded)
The number assigned to each visible proteoform row in the table. This sequential numbering does not change when you sort or filter the table.
Check box
Select this checkbox if you want to save the proteoforms in the selected rows with the protein sequence for a search.
To select or clear all of the checkboxes at once, select or clear the checkbox in the column header.
If you filter the table, the following occurs:
  • Clearing all checkboxes affects all of the original rows in the table, before any filtering.
  • Selecting all checkboxes affects only the filtered and currently visible rows.
Identification
Displays the protein sequence name, followed by an underscore, and then site(modification) for each modification separated by a comma.
For example, for "Protein_C4(Oxidation),N35(Deamidation)":
  • "Protein" represents the protein sequence name.
  • "C4" represents the site of the first modification.
  • "Oxidation" represents the first modification.
  • "N35" represents the site of the second modification.
  • "Deamidation" represents the second modification.
Modifications
Displays the modifications listed in the Identification column.
Sites
Displays the sites listed in the Identification column.
Num of Mods
Displays the number of modifications in the proteoform.
This number is within the range specified for the Number of Modifications per Proteoform parameter in the Site-Specific Variable Modifications for Top Down Analysis pane.
Mono. Mass
Displays the monoisotopic mass of the proteoform.
Avg. Mass
Displays the average mass of the proteoform.