In the Protein Sequence Editor, you can add variable modifications to the N-terminal, C-terminal, and side-chain residues in the protein sequence.
TIP
Upon installation, the BioPharma Finder application includes a default list of variable modifications. You can use the Define Modification List feature to customize the list visible in the Protein Sequence Editor and define a custom sublist for quick loading.
Prerequisites
- You are editing a protein sequence in the Protein Sequence Editor.
- The sequence has the "Peptide Mapping" category for Peptide Mapping Analysis or the "Intact Protein" category for Intact Mass Analysis.
Procedure
- In the Protein Sequence Editor, expand the Variable Modifications for Intact and Peptide Analysis pane on the right side of the page.
- At the top of the pane, in the Max # Modifications area, type the maximum number of modifications the application can apply to the sequence during Intact Mass Analysis (Intact Protein box) or Peptide Mapping Analysis (Peptide Mapping box).
- For Intact Mass Analysis, the maximum number of modifications is 10.
- For Peptide Mapping Analysis, the maximum number of modifications is 8.
- The maximum number of modifications does not include glycosylations.
IMPORTANT
Setting the "Max # of Modifications" parameter to a high value greatly increases the possible combinations of modifications in the sequence which might prolong job processing time.
- Next, add variable N-terminal, C-terminal, and side-chain modifications from the Modifications lists (left) to the Modifications Selected for Search lists (right):
- To assign a modification, select it from the Modifications list, and then select the Add button.
- To remove a modification, select it from the Modifications Selected for Search list, and then select the Remove button.
- To quickly assign all modifications in the default sublist, select the Load Default Mods button.
- Continue adding modifications from the Modifications list (left) to the Modifications Selected for Search lists (right) as necessary.
- Save the protein sequence.