The following table describes the parameters in the Residue Properties and Modifications dialog box in the Protein Sequence Editor.
Parameter | Description |
|---|---|
Residue Properties area | |
Residue | Displays the amino acid letter code of the selected residue. |
At | Displays the number of the chain and site location of the selected residue. |
Mono. Mass | Displays the monoisotopic mass of the residue. |
Avg. Mass | Displays the average mass of the residue. |
Side Chain/N-Terminal/C-Terminal Modification area | |
(Dropdown list) | Displays a list of the available side-chain, N-terminal, or C-terminal modifications. |
Mono. Mass | Displays the monoisotopic mass of the modification selected from the dropdown list. |
Avg. Mass | Displays the average mass of the modification selected from the dropdown list. |
Apply to All (checkbox) (Side Chain Modification area) | Applies the selected modification to all instances of the selected residue in the protein sequence. |
The following table describes the commands in the Residue Properties and Modifications dialog box in the Protein Sequence Editor.
Parameter | Description |
|---|---|
Residue Properties area | |
Residue | Displays the amino acid letter code of the selected residue. |
At | Displays the number of the chain and site location of the selected residue. |
Mono. Mass | Displays the monoisotopic mass of the residue. |
Avg. Mass | Displays the average mass of the residue. |
Side Chain/N-Terminal/C-Terminal Modification area | |
(Dropdown list) | Displays a list of the available side-chain, N-terminal, or C-terminal modifications. |
Mono. Mass | Displays the monoisotopic mass of the modification selected from the dropdown list. |
Avg. Mass | Displays the average mass of the modification selected from the dropdown list. |
Apply to All (checkbox) (Side Chain Modification area) | Applies the selected modification to all instances of the selected residue in the protein sequence. |