The following table describes the information in Disulfide Link Definitions table in the Protein Sequence Editor.
Column | Description |
|---|---|
Chain Number (1) | Displays the chain number of the first cysteine (C) residue in the disulfide link. |
Amino Acid Site Index (1) | Displays the chain-specific amino acid site of the first cysteine (C) residue in the disulfide link. |
Chain Number (2) | Displays the chain number of the second cysteine (C) residue in the disulfide link. |
Amino Acid Site Index (2) | Displays the chain-specific amino acid site of the second cysteine (C) residue in the disulfide link. |