The following table describes the parameters in the Modification Editor pane of the Protein Sequence Editor.
Parameter | Description |
---|---|
Modifications | Lists the default and custom N-terminal, C-terminal, and side-chain modifications available to assign to the protein sequence. |
Mono. Mass | Displays the monoisotopic mass of the selected modification. |
Avg. Mass | Displays the average mass of the selected modification. |
Residues (Side Chain area) | Displays the amino acid letter code(s) for the residue(s) associated with the selected modification. |
The following table describes the commands in the Modification Editor pane of the Protein Sequence Editor.
Button | Description |
---|---|
Add | Opens the Add New Modification dialog box to create a new custom modification. |
Delete | Deletes the selected custom modification from the Modifications lists in the Variable Modifications for Intact and Peptide Analysis and Modification Editor panes. |