The following table describes the information in the Sequence Manager table.
Column | Description |
---|---|
Name | Displays the name of the sequence. |
Sample Type | Displays the sample type of the sequence:
|
Category | Displays the category of the sequence. For proteins:
For oligonucleotides:
|
Last Modified Time | Displays the date and time that you last modified the sequence. |
Average Mass | Displays the average mass of the sequence. |
Monoisotopic Mass | Displays the monoisotopic mass of the sequence. |
Num. of Chains | Displays the number of chains in the sequence. |
Max. Num. of Modifications | Displays the maximum number of modifications for the sequence. This number is specified by a category-specific parameter in the Sequence Editor:
|
Protein Glycosylation/Oligo Terminal truncation | For protein sequences, displays the selected glycosylation in the protein sequence or None. For oligonucleotide sequences with the "Intact Deconvolution" category, displays Enabled (if Terminal Truncation Search is enabled) or None. |
Num. of Proteoforms | For protein sequences with the "Top Down" category, displays the total number of proteoforms generated. |
Variable Modifications | Displays all of the variable modifications in the sequence. |
Protein Static Modifications/Oligonucleotide Building Blocks | Displays all of the static modifications in protein sequences and the oligonucleotide building blocks in oligonucleotide sequences. |
Specificity | For oligonucleotide sequences with the "Intact Deconvolution" category and the Terminal Truncation Search enabled, displays the specificity for the sequence truncation:
|
Sequence Length | Displays the total number of amino acids (in a protein sequence) or nucleotide bases (in an oligonucleotide sequence). |
Description | Displays a description for each sequence (if applicable). |