The following table lists the O-linked glycans that the application supports for the O-glycan-specific search in peptide mapping analysis.
# | Glycan | Monoisotopic mass |
---|---|---|
1 | GalNAc | 203.079 |
2 | GalNAc-3G | 365.132 |
3 | GalNAc-3GnG | 568.212 |
4 | GalNAc-3SG | 656.228 |
5 | GalNAc-6GGn | 568.212 |
6 | GalNAc-6GGn-3G | 730.264 |
7 | GalNAc-6GGn-3GnG | 933.344 |
8 | GalNAc-6GGn-3SG | 1021.36 |
9 | GalNAc-6Gn | 406.159 |
10 | GalNAc-6Gn-3G | 568.212 |
11 | GalNAc-6Gn-3GnG | 771.291 |
12 | GalNAc-6Gn-3SG | 859.307 |
13 | GalNAc-6S | 494.175 |
14 | GalNAc-6S-3G | 656.228 |
15 | GalNAc-6S-3GnG | 859.307 |
16 | GalNAc-6S-3SG | 947.323 |