If you want to remove one or more filters the you apply to a table, follow the procedure below.
Procedure
  1. To remove a single filter:
  2. In the Results table on the Process and Review Page, place your cursor in the filter operand box of the column for which you want to remove the filter.
  3. Select the filter value that you previously typed or selected from the drop-down box and then press the DELETE key.
  4. The single filter is removed from the selected column and the Results table repopulates.
  5. To remove all filters in a table:
  6. In the Results table on the Process and Review Page, right-click anywhere in the table and select Filters > Clear All from the shortcut menu.
  7. All filters are removed from all table columns and the Results table repopulates.