For peptide mapping analysis or oligonucleotide analysis experiments, you can create, save, and apply custom Results table filters to a file to use in future experiments.
Prerequisites
- You have processed a peptide mapping analysis or oligonucleotide analysis experiment and are viewing the Results table on the Process and Review page.
Procedure
- In the Results table on the Process and Review page, select the down arrow (˅) in the column header of the column you want to filter and select (Custom) from the filter operand list.
- In the Custom Filter Selection dialog box, do the following for each condition that you want to add to the filter group:
- Select Add Condition.
- Select an operator from the Operator list
- Select an operand from the Operand list or type a specific text string or number.
- See Table filter operators and Table filter operands.
- By default, the application applies the AND operator to all of the conditions in the group.
Figure Custom Filter Selection dialog box - Select the space by the arrow icon (>) to the left of each condition row to select the row.
TIP
You can select contiguous rows by using the SHIFT key or noncontiguous rows using the CTRL key.
- To change from applying the AND operator for the group to applying the OR operator, and vice versa, select Toggle.
- A gray bar to the left of the condition rows indicates an AND group. This custom filter passes only if all of the conditions in the group pass.
- A blue bar to the left of the condition rows indicates an OR group. This custom filter passes if any of the conditions in the group passes.
- Select OK.
- The application applies the custom filter to the data in the selected filter column.
- Repeat this process to add additional column filters.