Procedure
- To copy all contents of a pane, do either of the following:
- In the title bar of the pane, select the Copy icon (if available) to copy all of the content in the pane.
- Right-click the pane and select Copy, Copy as Displayed, Copy per Global Settings, or Copy Data (if available).
- The Copy and Copy as Displayed commands copy the image in the pane as it is currently displayed.
- The Copy per Global Settings command copies the image at the set global dimensions.
- The Copy Data command copies the data in text format.
- To paste the copied content to a Microsoft Word™ (or similar text editor) file, select the Web Layout option at the bottom right of the main text editor window.
- In the text document, right-click and select the Keep Source Formatting option under Paste Options.
- To paste the contents in HTML format, right-click and select the Paste Special option.
- In the Paste Special dialog box, select the HTML option and then select OK.
NOTE
The content of some panes cannot be pasted into Microsoft PowerPoint™ or other presentation applications.