When you convert legacy data from a prior version of the BioPharma Finder application, the following occurs for sequences:
- The application migrates all sequences available on the Sequence Manager page of the legacy application.
- The Last Modified Time column on the Sequence Manager page displays the conversion time, not the original modified time.
- When you add a sequence to an experiment for intact mass analysis, the sequence name does not appear on the Load Results page. However, it appears on the Process and Review page in these areas:
- The Sequences Added to Experiment table under the Real Time Optimization > Identification subtab
- The Results table for a target sequence matching experiment
- The Matched Sequence pane
When you convert legacy data from the Protein Deconvolution application, the following occur for protein sequences:
- When you add a protein sequence to the method, the sequence name does not appear on the Load Results page or on the Process and Review page in the Sequences Added to Experiment table for real-time optimization. However, it appears in the Results table and the Matched Sequence pane.
- The BioPharma Finder application does not extract the sequence information from the results file and does not display this information on the Sequence Manager page.