The following table describes the commands on the Queue page.
Queue page commands
Commands
Description
Stop/Pause
(For Peptide Mapping Analysis, Oligonucleotide Analysis, and Top Down Analysis experiments) The Stop button cancels the processing of the currently running job and pauses the run queue.
(For Intact Mass Analysis experiments only) The Pause button completes the processing of the currently running job and then pauses the run queue.
Once you select the Stop/Pause button, the button changes to Run.
Run
The Run button resumes the processing of jobs in the run queue.
Once you select the Run button, the button changes to Stop or Pause.
Queue Manipulation
Contains sub-commands used to remove jobs from the run queue:
  • Remove Selected: Removes the selected job(s) from the queue
  • Remove Completed: Removes all completed jobs from the queue
  • Remove All: Removes all jobs from the queue
Open Results
Opens the experiment results for the selected job; transfers you to the Process and Review page.
This button is enabled for completed jobs only.
Open Report
(For Intact Mass Analysis experiments only) Opens the reports for the selected job; transfers you to the Reporting page.
This button is enabled for completed jobs only.