You can remove jobs in the run queue by using the Queue Manipulation button on the command bar to remove selected, completed, or all jobs.
Prerequisites
- You are viewing the Queue page.
Procedure
- (For selected jobs) On the Queue page, in the run queue table, select the row of the job(s) you want to remove.
TIP
To select multiple jobs simultaneously, select contiguous jobs using the SHIFT key or noncontiguous jobs using the CTRL key.
NOTE
You cannot remove jobs with a Running status from the run queue.
- On the command bar, select Queue Manipulation and then select of the following options:
- Remove Selected
- Remove Completed
- Remove All
- When prompted, select Yes in the confirmation box.
- The specified jobs are removed from the run queue table.
TIP
If you remove a job from the run queue, the experiment results can be still opened from the Load Results page.
Experiment results can be permanently deleted from the application on the Load Results page.