On the Queue page, you can start or suspend job processing using the Stop/Pause/Run and Queue Manipulation buttons on the command bar. You can also open experiment results and reports (for Intact Mass Analysis experiments).
| No. | Description | No. | Description |
|---|---|---|---|
1 | Queue page command bar | 4 | Open Results |
2 | Stop/Pause/Run button | 5 | Open Report |
3 | Queue Manipulation button (showing sub-commands) | 6 | Run queue table |
The following table describes the function of each command:
Command | Description | Result |
|---|---|---|
Stop | Appears on the Queue page for the Peptide Mapping Analysis, Oligonucleotide Analysis, and Top Down Analysis workflows only. Appears when an experiment is actively processing. Select Stop to stop a running experiment. | The experiment is cancelled and removed from the run queue. Once you select the Stop button, the button changes to Run. To rerun the job, you must recreate and resubmit the job as a new experiment. Other submitted jobs remain in the Submitted state in the run queue. |
Pause | Appears on the Queue page for the Intact Mass Analysis workflow only. Appears when an experiment is actively processing. Select Pause to complete the current experiment and then pause the run queue. | The current experiment completes processing and the queue is subsequently paused. Once you select the Pause button, the button changes to Run. To resume processing the remaining submitted experiments, select the Run button on the command bar. Other submitted jobs remain in the Submitted state in the run queue. You can submit new experiments for processing even when the run queue is paused. Any jobs you submit while the queue is paused will remain in the Submitted status until processing is resumed. |
Run | Appears on the Queue page for all workflows, but is active only when the Intact Mass Analysis queue has been paused or when a Top Down Analysis experiment is ready for processing. Select Run to process an experiment. | The experiment begins or resumes processing. Once you select the Run button, the button changes to Stop or Pause. |
Queue Manipulation | The Queue Manipulation button contains the following sub-commands:
When you select either the Cancel or Remove sub-commands, you must then select one of the following options:
| The selected (or all) experiments are deleted from the run queue. If you delete jobs from the run queue, the experiment results can be still opened from the Load Results page. Experiment results can be permanently deleted from the application on the Load Results page. In Ardia Platform mode, the Cancel or Remove action is recorded in the audit trail, but the draft experiment results are deleted from the Ardia Platform data repository. |
Open Results | This button is active for completed jobs only. Select Open Results to view the Process and Review page for the experiment. | Opens the results for the selected experiment; transfers you to the Process and Review page. |
Open Report | This button is active for completed jobs only. (For Intact Mass Analysis experiments only) Select Open Results to view the reports for the experiment. | Opens the reports for the selected experiment; transfers you to the Reporting page. |