Procedure
- On the BioPharma Finder Home page, in the Experiment Types pane, select the appropriate workflow link.
- The corresponding workflow page opens.
- In the Processing Method table, select the checkbox(es) for the processing method(s) you want to export.
- The Export Method button on the command bar becomes active.
Figure Active Export Method Button on the Intact Mass Analysis page - Select Export Method on the command bar.
- The Save As dialog box opens.
- In the Save As dialog box, navigate to the folder location where you want to save the method files (.
methodbpf
). TIP
For clarity, file names are appended according to their respective workflows.
- Select Save.
- A confirmation message appears.