The BioPharma Finder application requires one or more raw data files as input for every experiment, no matter the workflow. The raw data files contain your LC/MS experimental data and are processed by the application according to the processing method you select.
When you load multiple raw data files, you can assign a condition to each raw data file and select the reference condition for the experiment.
TIP
Copy the raw data files to your local computer for faster processing speed.
The application repeatedly accesses the raw data files during data processing and processing time might be very slow if the application accesses the raw data files through a network. The larger the number and file size of raw data files you load, the longer the application might take to process the results.