No matter the workflow you are using in BioPharma Finder, creating a new experiment involves the following tasks:
- Naming the experiment
- Loading the raw data files you want to analyze
- Selecting a target peptide, protein, or oligonucleotide sequence to apply to your experiment (if necessary)
- Specifying (and customizing, if needed) a processing method
- Processing your experiment in the run queue
- Viewing your experiment results
- Saving, exporting, and reporting your results as needed
The following sections describe the general procedures for completing these tasks in the BioPharma Finder application.
For detailed workflow-specific information about each of these tasks, refer to the corresponding chapters later in the user guide.