In local mode, you can generate and save point-in-time copies of the BioPharma Finder application.
TIP
The Snapshot feature is available only in local mode. In Ardia Platform mode, the application files are stored in a server and accessed from the Ardia Platform data repository.
Create a database Snapshot
Procedure
  1. On the BioPharma Finder Home page, select Help > Snapshot > Create.
  2. The Create a Snapshot dialog box appears.
  3. The application automatically names the Snapshot file as follows: BPF_version_MM_DD_YYYY_HH_mm_ss.DBbpf.
  4. (Optional) Select Browse to rename the Snapshot file, select a different location to save the Snapshot, and then select Save.
  5. You can create and save the Snapshot file to any location where you have write permissions. However, the location must have enough space to save the file. The application displays a message if there is not enough disk space for the file.
  6. Select Create.
  7. The Snapshot file (.DBbpf) is saved in the specified location and can be used for a point-in-time recovery.
Restore a database Snapshot
Procedure
  1. On the BioPharma Finder Home page, select Help > Snapshot > Restore.
  2. The Restore Snapshot dialog box appears.
  3. Select Browse to locate the Snapshot file (.DBbpf) that you want to restore, and then select Open.
  4. Select Restore.
  5. The selected Snapshot file replaces the current BioPharma Finder database with the Snapshot metadata.