TIP
Implement regular data backups to protect your data from unexpected loss and make it easy to revert back to a previous point in time if needed.
Although Thermo Scientific recommends using the imbedded Snapshot feature to perform a data backup, you can perform manual system backups in conjunction with this feature for added protection and flexibility.