When you open the Ardia Platform menu from the BioPharma Finder Home page, you can access the following commands to perform basic platform management tasks:
  • Connect: Connects the BioPharma Finder application to the Ardia Platform.
  • Disconnect: Disconnects the BioPharma Finder application from the Ardia Platform and restarts the application in local mode.
  • Sign Out: Signs out the current user from the Ardia Platform.
  • Register: Opens the Ardia Platform Registration dialog box.
  • Deregister: Deregisters the current user from the Ardia Platform.
  • Data Space Optimization: Removes temporary BioPharma Finder processing files to free up hard disk space.
Figure Ardia Platform menu when connected to the Ardia Platform
123456
No.DescriptionNo.Description
1
Connect to the Ardia Platform
4
Register with the Ardia Platform
2
Disconnect from the Ardia Platform
5
Deregister with the Ardia Platform
3
Sign Out from the Ardia Platform
6
Data Space Optimization submenu