IMPORTANT
In Ardia Platform mode, the Load Results page opens the Ardia Platform Data Explorer. For more information on using the Ardia Platform Data Explorer, refer to the Ardia Platform Help.
On the Load Results page, use the Experiment Management pane to manage and organize your saved experiment results.
TIP
You can access the Load Results page by selecting the appropriate tab on the navigation bar within a workflow.
By default, the Experiment Management pane is tabbed along the left side of the Load Results page. This pane contains two lists:
  1. The Master List displays all saved experiment results in a centralized list.
  2. The Working List displays the experiments that can be further organized into user-created folders or subfolders.
While all saved experiments are visible in the Master List at all times, experiments are removed from the Working List once they are added to a designated folder.
TIP
To toggle the display between the Master List and the Working List, expand the Experiment Management pane by selecting the double arrows in the left margin of the Load Results page and then select the list you want to display.