In the Compound Discoverer application, you can create and edit processing workflows (known as processing methods) within the study environment. If you need to create a new study, see Create a new study and a new analysis. In the Compound Discoverer application, you can open an existing study in multiple ways.
Procedure
- From the Compound Discoverer window, do any one of the following:
- From the Start Page, under What Would You Like to Do?, select Open Study, select a study file, and then select Open.
- From the application menu bar, select File > Open Study, select a study file, and then select Open.
- From the application toolbar, select Open an Existing Study, select a study file, and then select Open.
- The study opens and displays the Analysis Results page.