You can download the NIST 2023 MSMS libraries that are compatible with the Compound Discoverer application from the Life Sciences Mass Spectrometry (LSMS) Software Download and Licensing Portal.
Procedure
  1. Go to https://thermo.flexnetoperations.com.
  2. If you do not have access to the portal, register to create a new account as follows:
  3. On the LSMS Software Download and Licensing Portal, select the Register link.
  4. The Join Account page opens.
  5. Depending on your type of software, select either the Purchased Software tab or the Demo Software tab.
  6. Enter the required information, and then select Submit.
  7. Log in to your account.
  8. On the left side of the Life Sciences Mass Spectrometry Software Download and Licensing Portal, under Software and Services, select Product List .
  9. On the Product List page, select the Application-Compound Discoverer link.
  10. On the Product Information page, select the New Versions tab.
  11. Download the following files (these file names are subject to change):
  12. NIST 2023 mzVault format for CD.zip file.
  13. The Installing_and_Searching_NIST_2023_MSMS_Libraries.pdf.
  14. Extract the contents of the .zip file on a processing computer that has installations of the following applications:
  15. Compound Discoverer 3.0 or later
  16. (Optional) mzVault 2.3 or later