The Dionex ERD 500 contains components that are heat sensitive. The desalter should not be subjected to temperatures above 70°C during shipment, storage or operation.
Ensure the desalter is stored in a temperature controlled environment away from direct exposure to sunlight or other sources of heat. Do not store the desalter in a non-temperature controlled environment where temperatures exceed 70°C, such as a parked car.