Jump to main content
Tools and supplies
The following table lists the required tools and supplies for configuring the LC plumbing and EASY-Spray column connections. Not all parts are required.
NOTE
|
Item |
Part number |
|---|---|
|
One- and two-column setup |
|
|
A/B mixing/venting Tee, stainless steel, with three attached ferrules and nuts |
SC901 |
|
Two-column setup |
|
|
Acclaim™ PepMap100 Nano-Trap or other NanoViper™ equipped precolumn |
164705 |
|
PepMap™ Neo Trap Cartridge |
174500 |
|
Viper™ union |
6040.2304 |
|
UltiMate 3000 RSLCnano EASY-Spray Application Kit |
6720.0395 |