Configure the EASY-Spray source for NSI mode
Figure 1. NSI Source dialog box
NSI Source dialog box
Prerequisites
  • Configure the instrument using the Thermo Foundation Instrument Configuration.
Procedure
  1. Open the Tune application.
  2. Select Setup > NSI Source.
    NSI Source dialog box appears.
  3. Enter the following values:
    1. Spray Voltage (kV): 1.90
  4. Select OK.