Jump to main content
Figure 1. Two-column RSLCnano plumbing configuration.
Required tools and materials
Two-column RSLCnano plumbing configuration
CAUTION
-
EASY-Spray column
-
Viper union
-
20 μm ID nanoViper capillary
-
Precolumn
-
EASY-Spray column
-
Thermo Scientific Dionex™ UltiMate™ 3000 RSLCnano EASY-Spray Application Kit (P/N 6720.0395)
Prerequisites
-
Wear appropriate personal protective equipment (PPE), including a lab coat, safety glasses, and chemical-resistant gloves.
Procedure
- Connect the precolumn to ports 2 and 5 on one of the switching valves.
- Connect the 20 μm ID nanoViper capillary to port 4 on the same switching valve.
- Connect the fitting on the other end of the capillary to the Viper union. Tighten the fitting by hand.
- Connect the Viper union to the EASY-Spray column to complete the two-column plumbing configuration.