Temperature controller
The EASY-Spray Series ion source includes a temperature controller printed circuit board (PCB) that supplies power to and controls the temperature of the connected EASY-Spray column. The temperature controller includes an input power socket, a heater socket, a temperature control knob, and the Actual Temperature display.
NOTE
The input power socket (P-BUS) connects to the controller power supply or the EASY-Spray P-Bus Temp Control Cable. The heater socket connects to the heater cable on the EASY-Spray column.
The temperature control knob includes an OFF position and temperature settings from 30 °C to 60 °C in 5 °C increments. During operation, the Actual Temperature display shows the column temperature as it heats and stabilizes.
-
Actual Temperature display
-
Set Temperature knob
-
Heater input
-
Power input (P-BUS)