Install the USB camera
Install the USB camera and then use the DinoCaptureā„¢ application to view the tip of the emitter positioning tool and the spray from the EASY-Spray column.
Figure 1. USB camera installed on the ion source.
324
  1. Camera button
  2. Focusing wheel lock
  3. Focusing wheel (front faced)
  4. Camera mounting bracket
Prerequisites
  • Install the DinoCapture application on the data system computer before connecting the USB camera.
Procedure
  1. With the focusing wheel facing toward you, insert the USB camera into the camera opening.
  2. (Optional) Connect the USB camera to the USB extension cable.
  3. Connect the USB cable to a USB port on the data system computer.
    IMPORTANT
    Do not connect the USB camera to a USB hub.