The leak sensor is installed in the drip tray located near the bottom of the component panel. This error occurs when liquid accumulates in the drip tray.
Procedure
  1. Locate the source of the leak by visually inspecting the tubing, fittings, and components on the component panel. Also check for pressure changes; an increase or decrease in pressure may indicate a leak. For detailed troubleshooting of various types of leaks, see Liquid leaks.
  2. Tighten fittings, or replace tubing and fittings as required.
  3. Dry the drip tray and leak sensor thoroughly.
  4. NOTE
    After eliminating the source of a leak, always dry the drip tray and the leak sensor thoroughly. If the leak sensor is not dry, it will remain activated and will continue to report a leak to Chromeleon CDS.