The analytical pump head (P/N 22185-60213) consists of the entire analytical pump head assembly. For expanded views, see the following images.
Figure Analytical pump head components
Analytical pump head components
Item
Part number
Description
Quantity
1
22185-20211
Pump head block
1
2
22185-20212
Seal wash housing
2
3
22185-60214
Pump head rear plate
1
4
22185-98201
Piston seal
2
5
22185-98202
Sapphire piston
2
6
B51002818
Large O-ring
2
7
22185-98205
Ceramic washer
2
8
B51002819
Small O-ring
1
9
B51000563
Piston seal
2
Figure Analytical pump head components (view 2)
Analytical pump head components (view 2), continued
Item
Part number
Description
Quantity
1
22185-10201
Mounting plate
1
2
22185-10202
Face plate
1
3
22185-60213
Flangeless pump head
1
4
22185-60206
Transducer
1
5
22185-61008R
Pump patch PCB
1
6
22185-98200
Pump mechanism
1
7
045938
Screw to secure transducer block to mechanism
2
8
049649
Screw to secure face plate to mechanism
2
9
60-045686
Screw, M04 X0 70X006
1
10
60-050485
Screw for mounting plate
4
11
60-050490
Locking washer for mounting plate
4
12
00470-06315
Flat washer for M8 bolt
1
13
00671-01-00025
Locking washer for M8 bolt
1
14
C0619-01-00155
M8 bolt
1
15
22000-98001
PEEK fitting
4
16
22185-30201
Tubing
2
17
22185-60208
Outlet check valve
1
18
22185-60209
Inlet check valve
1
19
043276
Double-cone ferrule
4
20
B51003205
Label
1