Clean the check valves - Dionex Inuvion - Dionex Inuvion Operating Manual - User maintenance - Replace the pump check valves - Clean the check valves - Dionex Inuvion
Dionex Inuvion Operating Manual
- Product Tree
- Chromatography > Ion Chromatography > Dionex Inuvion
- Document Type
- User Guide
- Operating Manual
- Carefully remove the check valve cartridges from the valve housings.
- Place the check valve housings and cartridges in a beaker with methanol. Sonicate or agitate the parts for several minutes.
- Rinse each check valve housing and cartridge thoroughly with ASTM filtered, Type I (18.2 megohm-cm) deionized water that meets the specifications listed in Deionized water requirements for IC.
- To reinstall the check valves and complete the procedure, see Replace the pump check valves through Replace the pump check valves above.