Follow these guidelines to install and tighten a 10-32 fitting bolt (P/N 22000-98001) and 10-32 double-cone ferrule (P/N 043276).
For tightening requirements for IC PEEK Viper fittings, refer to the instructions provided with the fittings.
Procedure
  1. To install a 10-32 double-cone ferrule fitting, do the following:
  2. Install the fitting bolt and ferrule onto the tubing. Position the ferrule 1 to 2 mm (0.04 to 0.08 in) from the end of the tubing (see the following image).
  3. Figure Ferrule and fitting bolt placement for tubing connections
  4. Insert the tubing into the port until it stops.
  5. While maintaining pressure on the tubing to keep it in place in the port, tighten the fitting bolt finger-tight and then tighten the fitting further by following the instructions below.
  6. To tighten a 10-32 double-cone ferrule fitting, do the following:
  7. Use your fingers to tighten the fitting bolt as tight as you can. Then, use a wrench to tighten the fitting an additional three-quarter turn (270 degrees).
  8. If leaks occur, replace the fitting bolt, ferrule, and tubing.