To process samples manually, select operating parameters and commands from the Inuvion ePanel. Commands are executed as soon as they are entered.
Procedure
- Complete the instructions near the beginning of this chapter through Set system operating conditions to prepare the Dionex Inuvion IC System for operation and to prepare the sample for processing.
- Load the sample, using one of the following: a syringe, a vacuum syringe, or an autosampler.
- On the Inuvion ePanel, select the Autozero button.
- Inject the sample. The signal plot is displayed on the ePanel.
- Monitor the chromatogram. When the sample data has been collected, select the Monitor Baseline button on the Instrument toolbar above the ePanel Set.