For electrolytic eluent generation, or when manually preparing eluent and regenerant, use ASTM Type I (18.2 megohm-cm) filtered and deionized water that meets the specifications listed in the following table.
Contaminant | Specification |
|---|---|
Ions–Resistivity | >18.2 megohm-cm |
Organics–TOC | <10 ppb |
Iron/Transition Metals1) | <1 ppb |
Pyrogens | <0.03 (EU/mL) |
Particulates > 0.2 µm | <1 (units/mL) |
Colloids–Silica | <10 ppb |
Bacteria | <1 (CFU/mL) |