Procedure
- On the Inuvion ePanel, in the Valves area, select the Regen Pump button (see the following images).
Figure Hydrate the regenerant pump - This opens the Regen Pump Settingswindow.
- To start the regenerant pump, select the On button.
- Note that the default value for the pump flow rate is 3.00 mL/min, which is adjustable from 0 to 10 mL/min (the minimum rate is 0.10 mL/min) (see the following image). The flow rate is adjustable, based on the suppressor regenerant flow required for an application. The included regenerant pump tubing supports a maximum flow rate of 10 mL/min.
Figure Chromeleon CDS Regen Pump flow rate panel - After reaching the desired flow rate, close the Regen Pump Settings window. The regenerant pump is now ready for application testing.
NOTE
In instances where the conductivity background increases over time, check the regenerant flow rate. In general, the best practice is to confirm flow rate every other day; also check the flow rate when refilling regenerant and adjust as needed.