The Dionex Inuvion IC System can easily be moved by one person. Be sure to lift and carry the instrument by the base. Do not use the top rails as handles to lift or carry the instrument.
The Dionex Inuvion IC System can easily be moved by one person. Be sure to lift and carry the instrument by the base. Do not use the top rails as handles to lift or carry the instrument.