The Dionex Inuvion IC System is plumbed with the tubing and tubing assemblies listed below.
Tubing size and type
Color
Used for
0.125-mm (0.005-in) ID PEEK
(P/N 044221)
Red
Connection from pump pulse damper to pressure transducer
0.25-mm (0.010-in) ID PEEK
(P/N 042690)
Black
Connections between other system components
0.50-mm (0.020-in) ID PEEK
(P/N 042855)
Orange
Connection from injection port to injection valve
0.75-mm (0.030-in) ID PEEK
(P/N 044777)
Green
Connection from injection valve to waste
1.58-mm (0.062-in) ID PEEK
(P/N 014175)
Clear
Connection from pump to degas or eluent bottle; pump waste
25-µL sample loop (P/N 042857)
Orange
Connection between injection valve ports L (1) and L (4)
  • 10-32 fittings (P/N 22000-98001) and ferrules (P/N 043276) are used for most tubing connections.
  • 1/8-in fittings (P/N 052267) and ferrules (P/N 048949) are used for connections to the suppressor REGEN OUT port and the eluent bottle.