Daily maintenance - Dionex Inuvion - Dionex Inuvion Operating Manual - User maintenance - Daily maintenance - Routine user maintenance - Dionex Inuvion
Dionex Inuvion Operating Manual
- Product Tree
- Chromatography > Ion Chromatography > Dionex Inuvion
- Document Type
- User Guide
- Operating Manual
- Check the Dionex Inuvion IC System component panel for leaks or spills, especially around the drip tray (see Dionex Inuvion Core IC System component panel (with column holder and regenerant pump installed) and Instrument Overview.
- Wipe up spills. Isolate and repair leaks (see Liquid leaks). Rinse off any dried eluent with deionized water.
- Check the eluent bottle. When necessary, refill the eluent bottle with eluent for the Dionex Inuvion Core and Dionex Inuvion IC Systems. If the Dionex Inuvion IC System with RFIC is installed (or any system with RFIC capabilities), refill the eluent bottle with ASTM filtered, Type I (18.2 megohm-cm) deionized water that meets the specifications listed in Deionized water requirements for IC.
- Monitor the waste container and empty it as needed.