• To ensure pump reliability, replace the pump piston rinse seals, piston seals, and O-rings in the pump housing every 6 months, or whenever you replace the main piston seals (see Replace components in the analytical pump head assembly) for the Dionex Inuvion IC System pump
  • Replace the seal wash pump tubing.