Analytical pump and fluidics
Type
Serial dual-reciprocating pistons, microprocessor- controlled constant stroke, variable speed
Construction
Chemically inert, metal-free PEEK pump heads and flow paths compatible with aqueous eluents of pH 0 to 14
Pump operating pressure
0 to 35 MPa (0 to 5,000 psi)
Flow rate range
0.00 to 5.00 mL/min in 0.01 mL/min increments
Flow precision
<0.1%, typically
Flow accuracy
<0.1%, typically, at 13.8 MPa (2,000 psi) and 1.0 mL/min
Pressure ripple
<1%
Eluent on/off valve
Standard
Leak sensor
Optical, standard
Piston seal wash (optional)
Seal wash pump can be operated in continuous or intermittent mode when connected to rinse solution supply
Pressure alarm limits
Upper and lower limit pressure alarms can be set
Vacuum degas
Dionex Inuvion IC System: Standard user-adjustable vacuum level
Eluent bottles
Polypropylene bottles, 2 L and 4 L
Eluent bottle pressure (optional)
Supported by a digital controlled regulator with display; available for Dionex Core IC System and Dionex Inuvion IC System
Injection valve
6-port, 2-position Rheodyne valve, electronically activated
Columns supported
2-, 3-, 4-, and 5-mm ID; maximum length 250 mm analytical column with 5 mm guard column
Dionex eluent generation (Dionex Inuvion IC System)
Eluent types
KOH, MSA, or K2CO3
Eluent concentration range
KOH, MSA: Up to 100 mM
K2CO3: Up to 15 mM
Flow rate range
0.10 to 3.00 mL/min
Maximum operating pressure
35 MPa (5,000 psi)
Gradient profile
One-step linear
Column heater (optional)
Operating temperature range
30—60 °C (50—140 °F); settable within the software; minimum working range is 5 °C above ambient temperature
Temperature accuracy
±0.5 °C at sensor, at calibration points (30, 50 °C)
Suppressors and control (Dionex Inuvion Core IC System)
Chemical suppression
4 mm and 2 mm anion and cation suppressor types
Carbonic acid removal for anions
For use with hydroxide eluents: Dionex CRD 200
For use with carbonate eluents: Dionex CRD 300
Nonsuppressed chromatography
Yes, supported
Suppressor wear parts
Optional regenerant pump for chemical suppressor
Dynamic suppression capacity
Anions:
Dionex ACRS 500 (4 mm): 150 μeq/min
Dionex ACRS 500 (2 mm): 75 μeq/min
Cations:
Dionex CCRS 500 (4 mm): 75 μeq/min
Dionex CCRS 500 (2 mm): 37.5 μeq/min
Void volume
Dionex CRS 500 (4 mm): <50 μL
Dionex CRS 500 (2 mm): <15 μL
Chemical and electrolytic suppression
Anion and cation suppressor types, 4 mm and 2 mm
Suppressor regeneration mode
Electrolytic Suppressor: Recycle or external water mode
Chemical Suppressor: External regenerant
Current control range
Dionex DRS 600: 0—500 mA (4 mm) and 0—150 mA (2 mm) in 1 mA increments
Dionex ERD 500 Electrolytically Regenerated Desalter: 0—500 mA (4 mm) and 0—150 mA (2 mm) in 1 mA increments
Salt converter
Dionex SC-CERS 500 available in 4 mm and 2 mm versions
Carbonic acid removal for anions
For hydroxide eluents: Dionex CRD 200
For carbonate eluents: Dionex CRD 300
Nonsuppressed chromatography
Yes, supported
Suppressor wear parts
Chemical Suppression: Optional regenerant pump
Electrolytic Suppression: None
Suppressors and control (Dionex Inuvion IC System)
Chemical suppression
4 mm and 2 mm anion and cation suppressor types
Carbonic acid removal for anions
For use with hydroxide eluents: Dionex CRD 200
For use with carbonate eluents: Dionex CRD 300
Nonsuppressed chromatography
Yes, supported
Suppressor wear parts
Optional regenerant pump for chemical suppressor
Dynamic suppression capacity
Anions:
Dionex ACRS 500 (4 mm): 150 μeq/min
Dionex ACRS 500 (2 mm): 75 μeq/min
Dionex ADRS 600 (4 mm): 200 μeq/min
Dionex ADRS 600 (2 mm): 50 μeq/min
Dionex AERS 500e (4 mm): 200 μeq/min
Dionex AERS 500e (2 mm): 50 μeq/min
Dionex AERS 500 Carbonate (4 mm): 30 μeq/min
Dionex AERS 500 Carbonate (2 mm): 7.5 μeq/min
Cations:
Dionex CCRS 500 (4 mm): 75 μeq/min
Dionex CCRS 500 (2 mm): 37.5 μeq/min
Dionex CDRS 600 (4 mm): 100 μeq/min
Dionex CDRS 600 (2 mm): 35 μeq/min
Dionex CERS 500e (4 mm): 100 μeq/min
Dionex CERS 500e (2 mm): 35 μeq/min
Void volume
<50 μL for 4 mm Dionex DRS 600, Dionex ERS 500e, Dionex 500 Carbonate, and Dionex CRS 500 suppressors
<15 μL for 2 mm Dionex DRS 600, Dionex ERS 500e, Dionex ERS 500 Carbonate, and Dionex CRS 500 suppressors
Chemical and electrolytic suppression
Anion and cation suppressor types, 4 mm and 2 mm
Suppressor regeneration mode
Electrolytic Suppressor: Recycle or external water mode
Chemical Suppressor: External regenerant
Current control range
Dionex DRS 600: 0—500 mA (4 mm) and 0—150 mA (2 mm) in 1 mA increments
Dionex ERD 500 Electrolytically Regenerated Desalter: 0—500 mA (4 mm) and 0—150 mA (2 mm) in 1 mA increments
Salt converter
Dionex SC-CERS 500 available in 4 mm and 2 mm versions
Carbonic acid removal for anions
For hydroxide eluents: Dionex CRD 200
For carbonate eluents: Dionex CRD 300
Nonsuppressed chromatography
Yes, supported
Suppressor wear parts
Chemical Suppression: Optional regenerant pump
Electrolytic Suppression: None
Conductivity detector and flow cell
Type
Microprocessor-controlled digital signal processor
Cell drive
128 kHz square wave
Linearity
r2 ≥ 0.999%
Resolution
0.002 nS/cm
Full-scale output ranges
Digital input signal range: 0—18,000 μS/cm, with autoranging
Analog output signal range: 0—18,000 μS/cm
Temperature compensation
Fixed at 1.7% per 1 °C at cell temperature
Temperature range
Ambient +7 °C; 30—60 °C (86—122 ºF)
Cell electrodes
Passivated 316 stainless steel; compatible with methanesulfonic acid
Cell body
Chemically inert polymeric material
Cell volume
<1 μL
Heat exchanger
Inert, tortuous path for low axial dispersion
Maximum cell operating pressure
10 MPa (1,500 psi)
Data filter
Rise times from 0 to 10 s; user-selectable
Data collection rate
1—100 Hz, user-selectable
Autosampler
Automation using autosampler
Dionex AS-AP, Dionex AS-DV, Dionex AS-HV, or third-party autosampler
Sequential or simultaneous injection
Yes, depending on autosampler capabilities
Automated dilution
Yes, available with Dionex AS-AP
Dilution factor, Dionex AS-AP
1:1 to 1:1000
Dilution time, Dionex AS-AP
15 s with sample overlap
Inline sample degassing
Yes, optional with Dionex CRD 300 or Dionex CRD 200
Inline filtration
Yes, with Dionex AS-DV or inline filter
High automation flexibility
Conditionals using Chromeleon CDS and post-run features
Software
Chromeleon CDS supported on the following operating systems:
  • Windows 10 Enterprise and Pro
  • Windows 11 Enterprise and Pro
  • Autoconfiguration
  • Automated procedure wizard
  • System wellness and predictive performance
  • Data trending (numerical device parameters)
  • Virtual column simulator: (evaluation mode standard; isocratic and gradient optional)
  • Multi-vendor automation support of proprietary and third-party instruments (fully controls over 550 modules from more than 25 manufacturers, including GC, CE, HPLC, and MS)
  • Customizable system control panels
  • System trigger commands and conditionals
  • Data audit trail, system audit trail, and instrument audit trail
  • Multiple network control and network failure protection (optional)
  • System calibration storage (factory, present, and previous; completely user-selectable)
  • Automated system qualification (detailed, comprehensive qualification reports)
  • Dual sequence view in the studio
Physical
Use environment
Indoor, dry location
Operating temperature
4—40 °C (40—104 °F)
Operating humidity range
20—80% relative humidity, noncondensing
Altitude
< 2,000 m (6,562 ft) AMSL
Pollution degree
II
Power requirements
100—240 Vac, 50 or 60 Hz autoranging, 5 A
Control modes
Full control through Chromeleon CDS; alternative control through TTL or relay closures (one relay output, two TTL outputs, two assignable TTL inputs)
USB communication protocol
One USB input; three USB outputs
Shipping dimensions (h × w × d)
66.1 × 29.2 × 43.3 cm (26.0 × 11.5 × 17.0 in)
Weight
Dionex Inuvion Core IC System: 15.3 kg (33.7 lbs)
Dionex Inuvion IC System: 16.2 kg (36 lbs)