Data from manual processing is saved in the manual sequence under the instrument folder in the local data vault.
Procedure
  1. Select the manual folder and select File > Save As.
  2. Enter a new name for the sequence.
  3. Select the Save raw data checkbox.
  4. Select Save.