Procedure
  1. Disconnect the waste line from the port on the injection valve and replace it with a piece of PEEK or PTFE (polytetrafluoroethylene) tubing that is 25 to 30 cm (10 to 12 in) long.
  2. Place the free end of the PEEK or PTFE line into the sample.
  3. Verify that the injection valve is in the Load position.
  4. Insert the 10-cc syringe (P/N 079803) provided in the Dionex Inuvion IC System Ship Kit (PN 22185-62000) into the injection port on the front door and pull out the plunger to draw the sample into the injection valve.
  5. Switch the injection valve to the Inject position (see Inject samples).