Load samples with a vacuum syringe - Dionex Inuvion - Dionex Inuvion Operating Manual - Operation and user setup - Load samples with a vacuum syringe - Preparing samples - Loading and injecting samples - Setting up the autosampler - Dionex Inuvion
Dionex Inuvion Operating Manual
- Product Tree
- Chromatography > Ion Chromatography > Dionex Inuvion
- Document Type
- User Guide
- Operating Manual
- Disconnect the waste line from port W (6) on the injection valve and replace it with a piece of PEEK or PTFE (polytetrafluoroethylene) tubing that is 25 to 30 cm (10 to 12 in) long.
- Place the free end of the PEEK or PTFE line into the sample.
- Verify that the injection valve is in the Load position.
- Insert the 10-cc syringe (P/N 079803) provided in the Dionex Inuvion IC System Ship Kit (PN 22185-62000) into the injection port on the front door and pull out the plunger to draw the sample into the injection valve.
- Switch the injection valve to the Inject position (see Inject samples).