Load samples with a syringe - Dionex Inuvion - Dionex Inuvion Operating Manual - Load samples with a syringe - Operation and user setup - Preparing samples - Loading and injecting samples - Setting up the autosampler - Dionex Inuvion
Chromatography > Ion Chromatography > Dionex Inuvion
Document Type
User Guide
Operating Manual
Procedure
Verify that the injection port on the Dionex Inuvion IC System front panel is connected to sample port S (5) on the injection valve (see the following image).
Figure Injection valve plumbing connections
Fill the 10-cc syringe (P/N 079803) provided in the system Ship Kit (PN 22185-62000) with a calibration standard or sample.
Insert the syringe into the injection port on the component panel.
Verify that the injection valve is in the Load position.
Overfill the sample loop with several sample loop volumes. Excess sample will exit through the injection valve waste line.
Leave the syringe in the port and switch the injection valve to the Inject position.