The Dionex Inuvion IC System does not require pressurized bottles. However, if the eluent is manually degassed or is sensitive to contamination, Thermo Fisher Scientific recommends pressurizing the bottle with helium or nitrogen.
An eluent monitor module can be added to the instrument configuration to continuously monitor the eluent volume and alert you when the level is below the user-specified threshold. Each eluent monitor can support one bottle. You can monitor up to three bottles, using the appropriate kit(s):
- Liquid Level Sensing Kit for 2-liter bottles (P/N 22815-62707)
- Liquid Level Sensing Kit for 4-liter bottles (P/N 22815-62708)
For more information, see Install the eluent monitor.
The digital pressure regulator (PN 22185-62706) required for pressurizing eluent bottle(s) must be ordered separately. Do not directly pressurize an eluent bottle from a gas source.
WARNING
Risk of bottle rupture or leakage: If pressurized bottles are required, use the Thermo Scientific Digital Pressure Regulator (P/N 22185-62706).
WARNING
Risk of bottle rupture or leakage: Do not pressurize eluent bottles above 10 psi (69 kPa).
Task | Procedure |
---|---|
Filter the deionized water | Filtering removes small particulates in deionized water that may contaminate the pump check valves and cause erratic flow rates or loss of prime. Filtration is advised, but not required, when using a Dionex Inuvion IC RFIC System. Procedure
|
Fill the eluent bottle | Fill the bottle with ASTM filtered, Type I (18.2 megohm-cm) deionized water that meets the specifications in Deionized water requirements for IC. |
Connect the eluent bottle | If it is not already connected, connect the ELUENT BOTTLE OUT line from the bottle cap to the ELUENT IN line, which extends from the tubing chase on the top cover of the Dionex Inuvion IC System. |