Possible cause | Solution |
---|---|
Background not suppressed by suppressor | Verify that the suppressor is turned on and that the current is set to the correct value. For additional troubleshooting guidance, refer to the suppressor manual. |
Sample concentration too high | Dilute the sample (see Dilute samples). |
Wrong eluent | Check the column manual to verify that you are using the correct eluent for your application. |
Background conductivity not offset from conductivity reading | Procedure
|